siRNA Details
| Gene Symbol: Hsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768238.1 | XP_008766460.1 | 5 | 2168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsf5| Gene Symbol: Hsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768238.1 | XP_008766460.1 | 5 | 2168 | ||