siRNA Details
| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271277.1 | NP_001258206.1 | 1 | 38 | ||
| XM_008768256.1 | XP_008766478.1 | 1 | 36 | |||
| GenBank mRNA | FQ216780.1 | - | 1 | 38 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcap| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271277.1 | NP_001258206.1 | 1 | 38 | ||
| XM_008768256.1 | XP_008766478.1 | 1 | 36 | |||
| GenBank mRNA | FQ216780.1 | - | 1 | 38 | ||