siRNA Details
| Gene Symbol: Myl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109495.1 | NP_001102965.1 | 4, 5 | 556 | ||
| GenBank mRNA | FQ217482.1 | - | 2, 3 | 409 | ||
| X51531.1 | - | 4, 5 | 535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myl4| Gene Symbol: Myl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109495.1 | NP_001102965.1 | 4, 5 | 556 | ||
| GenBank mRNA | FQ217482.1 | - | 2, 3 | 409 | ||
| X51531.1 | - | 4, 5 | 535 | |||