siRNA Details
| Gene Symbol: Nt5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271189.1 | NP_001258118.1 | 3 | 335 | ||
| GenBank mRNA | FQ227491.1 | - | 3 | 346 | ||
| FQ233678.1 | - | 3 | 335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nt5c| Gene Symbol: Nt5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271189.1 | NP_001258118.1 | 3 | 335 | ||
| GenBank mRNA | FQ227491.1 | - | 3 | 346 | ||
| FQ233678.1 | - | 3 | 335 | |||