siRNA Details
| Gene Symbol: Rnf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768460.1 | XP_008766682.1 | 2, 3 | 279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf157| Gene Symbol: Rnf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768460.1 | XP_008766682.1 | 2, 3 | 279 | ||