siRNA Details
| Gene Symbol: Tmem235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247851.2 | XP_006247913.1 | 6 | 2935 | ||
| XM_006247852.2 | XP_006247914.1 | 6 | 1875 | |||
| XM_008768444.1 | XP_008766666.1 | 5 | 3073 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem235| Gene Symbol: Tmem235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247851.2 | XP_006247913.1 | 6 | 2935 | ||
| XM_006247852.2 | XP_006247914.1 | 6 | 1875 | |||
| XM_008768444.1 | XP_008766666.1 | 5 | 3073 | |||