siRNA Details
| Gene Symbol: Aspscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001076577.5 | XP_001076577.3 | 12 | 1845 | ||
| GenBank mRNA | BC162026.1 | - | 12 | 1406 | 9099544 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aspscr1| Gene Symbol: Aspscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001076577.5 | XP_001076577.3 | 12 | 1845 | ||
| GenBank mRNA | BC162026.1 | - | 12 | 1406 | 9099544 | |