siRNA Details
| Gene Symbol: Pabpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110366.2 | NP_001103836.1 | 1 | 500 | ||
| XM_008773376.1 | XP_008771598.1 | 2 | 891 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc5| Gene Symbol: Pabpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110366.2 | NP_001103836.1 | 1 | 500 | ||
| XM_008773376.1 | XP_008771598.1 | 2 | 891 | |||