siRNA Details
| Gene Symbol: Ckmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127652.1 | NP_001121124.1 | 1023 | |||
| GenBank mRNA | BC158762.1 | - | 1023 | 7105696 | ||
| FQ215460.1 | - | 5 | 675 | |||
| FQ216641.1 | - | 7 | 1064 | |||
| FQ224910.1 | - | 7 | 1062 | |||
| X59736.1 | - | 7 | 1051 | |||

Ckmt2
Genomic Map