siRNA Details
| Gene Symbol: Ckmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127652.1 | NP_001121124.1 | 1192 | |||
| GenBank mRNA | BC158762.1 | - | 1192 | 7105696 | ||
| FQ215460.1 | - | 6, 7 | 844 | |||
| FQ216242.1 | - | 6, 7 | 1020 | |||
| FQ216641.1 | - | 8, 9 | 1233 | |||
| FQ224910.1 | - | 8, 9 | 1231 | |||
| X59736.1 | - | 8, 9 | 1220 | |||

Ckmt2
Genomic Map