siRNA Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128083.1 | NP_001121555.1 | 6 | 2657 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim8| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128083.1 | NP_001121555.1 | 6 | 2657 | ||