siRNA Details
Gene Symbol: Ryr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191043.1 | NP_001177972.1 | 13, 14 | 1696 | ||
NM_032078.2 | NP_114467.1 | 13, 14 | 1602 | |||
GenBank mRNA | AB204523.1 | - | 13, 14 | 1696 | ||
BC167757.1 | - | 13, 14 | 1987 | |||
EU346200.2 | - | 13, 14 | 1602 |
