siRNA Details
| Gene Symbol: Bag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128195.1 | NP_001121667.1 | 3 | 303 | ||
| GenBank mRNA | BC158711.1 | - | 3 | 303 | 9035120 | |
| FQ228417.1 | - | 3 | 327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bag2| Gene Symbol: Bag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128195.1 | NP_001121667.1 | 3 | 303 | ||
| GenBank mRNA | BC158711.1 | - | 3 | 303 | 9035120 | |
| FQ228417.1 | - | 3 | 327 | |||