siRNA Details
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110494.1 | NP_001103964.1 | 12 | 1370 | ||
XM_006255816.2 | XP_006255878.1 | 12 | 1388 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110494.1 | NP_001103964.1 | 12 | 1370 | ||
XM_006255816.2 | XP_006255878.1 | 12 | 1388 |