siRNA Details
| Gene Symbol: Slc2a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101020.1 | NP_001094490.1 | 1331 | |||
| GenBank mRNA | EF501821.1 | - | 1331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a7| Gene Symbol: Slc2a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101020.1 | NP_001094490.1 | 1331 | |||
| GenBank mRNA | EF501821.1 | - | 1331 | |||