siRNA Details
| Gene Symbol: Maz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110319.1 | NP_001103789.1 | 4 | 1171 | ||
| GenBank mRNA | EF059745.1 | - | 4 | 1171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maz| Gene Symbol: Maz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110319.1 | NP_001103789.1 | 4 | 1171 | ||
| GenBank mRNA | EF059745.1 | - | 4 | 1171 | ||