siRNA Details
| Gene Symbol: LOC100912347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243414.2 | XP_006243476.1 | 4 | 2178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100912347| Gene Symbol: LOC100912347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243414.2 | XP_006243476.1 | 4 | 2178 | ||