siRNA Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047913.2 | NP_001041378.2 | 6 | 960 | ||
| XM_006234887.2 | XP_006234949.1 | 7 | 1032 | |||
| GenBank mRNA | BC158559.1 | - | 6 | 960 | 7456444 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sqrdl| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047913.2 | NP_001041378.2 | 6 | 960 | ||
| XM_006234887.2 | XP_006234949.1 | 7 | 1032 | |||
| GenBank mRNA | BC158559.1 | - | 6 | 960 | 7456444 | |