siRNA Details
Gene Symbol: Slc35g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127658.1 | NP_001121130.1 | 2 | 114 | ||
GenBank mRNA | BC159402.1 | - | 2 | 114 | 7457005 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc35g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127658.1 | NP_001121130.1 | 2 | 114 | ||
GenBank mRNA | BC159402.1 | - | 2 | 114 | 7457005 |