siRNA Details
| Gene Symbol: Akr1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031000.3 | NP_112262.1 | 6, 7 | 815 | ||
| GenBank mRNA | BC059133.1 | - | 6, 7 | 1094 | 6918695 | |
| D10854.1 | - | 6, 7 | 815 | |||
| FQ212523.1 | - | 6, 7 | 1058 | |||
| FQ212586.1 | - | 6, 7 | 1055 | |||
| FQ212779.1 | - | 6, 7 | 1090 | |||
| FQ212919.1 | - | 5, 6 | 765 | |||
| FQ213220.1 | - | 5, 6 | 762 | |||
| FQ213467.1 | - | 6, 7 | 1090 | |||
| FQ213483.1 | - | 6, 7 | 1086 | |||
| FQ213491.1 | - | 6, 7 | 1088 | |||
| FQ213579.1 | - | 6, 7 | 1058 | |||
| FQ213776.1 | - | 6, 7 | 1057 | |||
| FQ214173.1 | - | 6, 7 | 1058 | |||
| FQ214441.1 | - | 6, 7 | 834 | |||
| FQ214642.1 | - | 1071 | ||||
| FQ214700.1 | - | 6, 7 | 1031 | |||
| FQ215772.1 | - | 6, 7 | 1058 | |||
| FQ218383.1 | - | 6, 7 | 1058 | |||
| FQ218721.1 | - | 6, 7 | 1057 | |||
| FQ218729.1 | - | 6, 7 | 1057 | |||
| FQ220055.1 | - | 4, 5 | 658 | |||
| FQ226221.1 | - | 6, 7 | 1090 | |||
| FQ228131.1 | - | 6, 7 | 1091 | |||
| FQ228132.1 | - | 6, 7 | 1090 | |||
| FQ228842.1 | - | 6, 7 | 1086 | |||
| FQ229180.1 | - | 5, 6 | 729 | |||
| FQ233156.1 | - | 6, 7 | 915 | |||

Akr1a1
Genomic Map