siRNA Details
| Gene Symbol: Arhgef11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023982.1 | NP_076472.1 | 5 | 221 | ||
| XM_006232759.1 | XP_006232821.1 | 5 | 1363 | |||
| XM_006232766.1 | XP_006232828.1 | 3 | 1315 | |||
| XM_008761214.1 | XP_008759436.1 | 3 | 1302 | |||
| XM_008761215.1 | XP_008759437.1 | 3 | 1302 | |||
| XM_008761216.1 | XP_008759438.1 | 3 | 1299 | |||
| XM_008761217.1 | XP_008759439.1 | 3 | 1299 | |||
| XM_008761218.1 | XP_008759440.1 | 4 | 2276 | |||
| XM_008761219.1 | XP_008759441.1 | 3 | 1302 | |||
| XM_008761220.1 | XP_008759442.1 | 3 | 1299 | |||
| XM_008761221.1 | XP_008759443.1 | 3 | 2261 | |||
| XM_008761222.1 | XP_008759444.1 | 3 | 1302 | |||
| XM_008761223.1 | XP_008759445.1 | 3 | 1302 | |||
| XM_008761224.1 | XP_008759446.1 | 3 | 1299 | |||
| XM_008761225.1 | XP_008759447.1 | 3 | 1302 | |||
| XM_008761226.1 | XP_008759448.1 | 4 | 2276 | |||
| GenBank mRNA | AF225961.1 | - | 5 | 221 | ||

Arhgef11
Genomic Map