siRNA Details
| Gene Symbol: Srebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276707.1 | NP_001263636.1 | 14 | 2552 | ||
| NM_001276708.1 | NP_001263637.1 | 14 | 2439 | |||
| XM_008767879.1 | XP_008766101.1 | 14 | 2692 | |||
| GenBank mRNA | AF286469.2 | - | 14 | 2406 | ||
| AF286470.2 | - | 14 | 2406 | |||

Srebf1
Genomic Map