siRNA Details
| Gene Symbol: Tas2r107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023995.1 | NP_076485.1 | 1 | 174 | ||
| GenBank mRNA | AF227142.1 | - | 1 | 174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r107| Gene Symbol: Tas2r107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023995.1 | NP_076485.1 | 1 | 174 | ||
| GenBank mRNA | AF227142.1 | - | 1 | 174 | ||