siRNA Details
| Gene Symbol: Fgfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024146.1 | NP_077060.1 | 17 | 2326 | ||
| GenBank mRNA | D12498.1 | - | 17 | 2326 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgfr1| Gene Symbol: Fgfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024146.1 | NP_077060.1 | 17 | 2326 | ||
| GenBank mRNA | D12498.1 | - | 17 | 2326 | ||