siRNA Details
| Gene Symbol: Evl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024147.1 | NP_077061.1 | 2, 3 | 394 | ||
| XM_008764850.1 | XP_008763072.1 | 2, 3 | 343 | |||
| XM_008764851.1 | XP_008763073.1 | 2, 3 | 2226 | |||
| XM_008764852.1 | XP_008763074.1 | 3, 4 | 2191 | |||
| XM_008764853.1 | XP_008763075.1 | 3, 4 | 2191 | |||
| XM_008764854.1 | XP_008763076.1 | 3, 4 | 2156 | |||
| XM_008764855.1 | XP_008763077.1 | 2, 3 | 343 | |||
| XM_008764856.1 | XP_008763078.1 | 2, 3 | 494 | |||
| XM_008764857.1 | XP_008763079.1 | 2, 3 | 321 | |||
| GenBank mRNA | U70211.1 | - | 2, 3 | 394 | ||

Evl
Genomic Map