siRNA Details
| Gene Symbol: Arf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024152.2 | NP_077066.1 | 1 | 230 | ||
| GenBank mRNA | BC091146.1 | - | 1 | 230 | 7309902 | |
| L12385.1 | - | 1 | 219 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arf6| Gene Symbol: Arf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024152.2 | NP_077066.1 | 1 | 230 | ||
| GenBank mRNA | BC091146.1 | - | 1 | 230 | 7309902 | |
| L12385.1 | - | 1 | 219 | |||