siRNA Details
| Gene Symbol: Dab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024159.1 | NP_077073.1 | 10 | 1445 | ||
| GenBank mRNA | BC097314.1 | - | 10 | 1480 | 7444786 | |
| FQ224185.1 | - | 1101 | ||||
| U95177.1 | - | 10 | 1445 | |||
| U95178.1 | - | 9 | 791 | |||

Dab2
Genomic Map