siRNA Details
Gene Symbol: Begain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111115.1 | NP_001104585.1 | 7 | 1748 | ||
NM_024163.2 | NP_077077.1 | 7 | 1768 | |||
XM_008764858.1 | XP_008763080.1 | 8 | 2166 | |||
XM_008764859.1 | XP_008763081.1 | 7 | 2076 | |||
XM_008764860.1 | XP_008763082.1 | 7 | 2278 | |||
GenBank mRNA | AF064868.1 | - | 7 | 1803 | ||
AF064869.1 | - | 7 | 1761 |
