siRNA Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169137.2 | NP_001162608.1 | 7, 8 | 2050 | ||
| NM_024367.1 | NP_077343.1 | 6, 7 | 1714 | |||
| XM_006250819.2 | XP_006250881.1 | 7, 8 | 2559 | |||
| GenBank mRNA | X78689.1 | - | 6, 7 | 1781 | ||

Epha5
Genomic Map