siRNA Details
| Gene Symbol: Gdf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024375.1 | NP_077351.1 | 1613 | |||
| GenBank mRNA | D49494.1 | - | 1613 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf10| Gene Symbol: Gdf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024375.1 | NP_077351.1 | 1613 | |||
| GenBank mRNA | D49494.1 | - | 1613 | |||