siRNA Details
| Gene Symbol: Hes5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024383.1 | NP_077359.1 | 1 | 85 | ||
| GenBank mRNA | D12516.1 | - | 1 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hes5| Gene Symbol: Hes5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024383.1 | NP_077359.1 | 1 | 85 | ||
| GenBank mRNA | D12516.1 | - | 1 | 85 | ||