siRNA Details
| Gene Symbol: Thoc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024384.2 | NP_077360.2 | 4 | 494 | ||
| GenBank mRNA | BC079149.1 | - | 4 | 494 | 7128541 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thoc6| Gene Symbol: Thoc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024384.2 | NP_077360.2 | 4 | 494 | ||
| GenBank mRNA | BC079149.1 | - | 4 | 494 | 7128541 | |