siRNA Details
| Gene Symbol: Hmox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277073.1 | NP_001264002.1 | 6 | 1022 | ||
| NM_024387.2 | NP_077363.1 | 6 | 1066 | |||
| XM_006245827.2 | XP_006245889.1 | 6 | 1244 | |||
| XM_008767500.1 | XP_008765722.1 | 5 | 1973 | |||
| GenBank mRNA | BC062061.1 | - | 6 | 981 | 5599345 | |
| FM110439.1 | - | 3 | 327 | |||
| FM139649.1 | - | 3 | 488 | |||
| FQ225862.1 | - | 6 | 1020 | |||
| J05405.1 | - | 6 | 1058 | |||

Hmox2
Genomic Map