siRNA Details
| Gene Symbol: Hmox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277073.1 | NP_001264002.1 | 4 | 767 | ||
| NM_024387.2 | NP_077363.1 | 4 | 811 | |||
| XM_006245827.2 | XP_006245889.1 | 4 | 989 | |||
| XM_008767500.1 | XP_008765722.1 | 3 | 1718 | |||
| GenBank mRNA | BC062061.1 | - | 4 | 726 | 5599345 | |
| FM110439.1 | - | 1 | 72 | |||
| FM139649.1 | - | 1 | 233 | |||
| FQ225862.1 | - | 4 | 765 | |||
| J05405.1 | - | 4 | 803 | |||

Hmox2
Genomic Map