siRNA Details
| Gene Symbol: Abi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024397.2 | NP_077373.2 | 10, 11 | 1324 | ||
| XM_006254353.2 | XP_006254415.1 | 10, 11 | 1392 | |||
| XM_006254354.2 | XP_006254416.1 | 9, 10 | 1379 | |||
| XM_006254355.2 | XP_006254417.1 | 9, 10 | 1376 | |||
| XM_006254356.2 | XP_006254418.1 | 9, 10 | 1307 | |||
| XM_006254357.2 | XP_006254419.1 | 8, 9 | 1289 | |||
| XM_006254358.2 | XP_006254420.1 | 8, 9 | 1130 | |||
| XM_006254359.2 | XP_006254421.1 | 7, 8 | 1115 | |||
| XM_006254360.2 | XP_006254422.1 | 9, 10 | 1322 | |||
| GenBank mRNA | AF176784.1 | - | 9, 10 | 1246 | ||
| BC129092.1 | - | 10, 11 | 1280 | 7192971 | ||
| CB725283.1 | - | 3, 4 | 461 | |||

Abi1
Genomic Map