siRNA Details
| Gene Symbol: Ube2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031001.2 | NP_112263.1 | 1 | 381 | ||
| GenBank mRNA | BC078808.1 | - | 1 | 381 | 7113229 | |
| U56407.1 | - | 1 | 272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2d4| Gene Symbol: Ube2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031001.2 | NP_112263.1 | 1 | 381 | ||
| GenBank mRNA | BC078808.1 | - | 1 | 381 | 7113229 | |
| U56407.1 | - | 1 | 272 | |||