siRNA Details
| Gene Symbol: Alcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031753.1 | NP_113941.1 | 5 | 679 | ||
| GenBank mRNA | AB008538.1 | - | 5 | 679 | ||
| BC061970.1 | - | 5 | 688 | 5621856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alcam| Gene Symbol: Alcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031753.1 | NP_113941.1 | 5 | 679 | ||
| GenBank mRNA | AB008538.1 | - | 5 | 679 | ||
| BC061970.1 | - | 5 | 688 | 5621856 | ||