siRNA Details
| Gene Symbol: Grpel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024487.4 | NP_077813.1 | 3, 4 | 330 | ||
| XM_006251142.2 | XP_006251204.1 | 4, 5 | 523 | |||
| GenBank mRNA | BC097312.1 | - | 3, 4 | 328 | 7387647 | |
| FQ219742.1 | - | 4, 5 | 511 | |||
| FQ220295.1 | - | 3, 4 | 321 | |||
| U62940.1 | - | 3, 4 | 331 | |||

Grpel1
Genomic Map