siRNA Details
| Gene Symbol: Hnrnpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134760.1 | NP_001128232.1 | 9 | 1547 | ||
| NM_032619.2 | NP_116008.2 | 10 | 1436 | |||
| XM_006228691.2 | XP_006228753.1 | 9 | 1691 | |||
| XM_006228692.2 | XP_006228754.1 | 10 | 1580 | |||
| GenBank mRNA | AB260892.1 | - | 3 | 413 | ||
| AF260436.1 | - | 1 | 100 | |||
| BC086392.1 | - | 6 | 661 | |||
| BC169048.1 | - | 10 | 1391 | |||

Hnrnpl
Genomic Map