siRNA Details
| Gene Symbol: Serp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030835.2 | NP_110462.1 | 1 | 385 | ||
| GenBank mRNA | AB018546.1 | - | 1 | 270 | ||
| AF100470.1 | - | 1 | 39 | |||
| AJ238236.1 | - | 1 | 45 | |||
| BC061854.1 | - | 1 | 385 | 5621665 | ||

Serp1
Genomic Map