siRNA Details
Gene Symbol: Serp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030835.2 | NP_110462.1 | 2 | 495 | ||
GenBank mRNA | AB018546.1 | - | 2 | 380 | ||
AF100470.1 | - | 2 | 149 | |||
AJ238236.1 | - | 2 | 155 | |||
BC061854.1 | - | 2 | 495 | 5621665 |
