siRNA Details
Gene Symbol: Trim23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760683.1 | XP_008758905.1 | 9 | 1362 | ||
XM_008760684.1 | XP_008758906.1 | 9 | 1263 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760683.1 | XP_008758905.1 | 9 | 1362 | ||
XM_008760684.1 | XP_008758906.1 | 9 | 1263 |