siRNA Details
Gene Symbol: Trim23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100637.1 | NP_001094107.1 | 10 | 1554 | ||
XM_008760683.1 | XP_008758905.1 | 11 | 1635 | |||
XM_008760684.1 | XP_008758906.1 | 11 | 1536 | |||
GenBank mRNA | CK470479.1 | - | 1 | 56 | ||
L04760.1 | - | 10 | 1405 |
