siRNA Details
| Gene Symbol: Itga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030842.1 | NP_110469.1 | 1 | 91 | ||
| XM_006240800.2 | XP_006240862.1 | 1 | 269 | |||
| GenBank mRNA | BC088846.1 | - | 1 | 91 | 7313230 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga7| Gene Symbol: Itga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030842.1 | NP_110469.1 | 1 | 91 | ||
| XM_006240800.2 | XP_006240862.1 | 1 | 269 | |||
| GenBank mRNA | BC088846.1 | - | 1 | 91 | 7313230 | |