siRNA Details
| Gene Symbol: Cxcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030845.1 | NP_110472.1 | 2 | 195 | ||
| GenBank mRNA | D11444.1 | - | 2 | 180 | ||
| M86536.1 | - | 2 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl1| Gene Symbol: Cxcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030845.1 | NP_110472.1 | 2 | 195 | ||
| GenBank mRNA | D11444.1 | - | 2 | 180 | ||
| M86536.1 | - | 2 | 195 | |||