siRNA Details
| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030849.1 | NP_110476.1 | 8 | 1070 | ||
| GenBank mRNA | D17667.1 | - | 7 | 845 | ||
| D38082.1 | - | 7 | 927 | |||
| S75359.1 | - | 8 | 1070 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmpr1a| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030849.1 | NP_110476.1 | 8 | 1070 | ||
| GenBank mRNA | D17667.1 | - | 7 | 845 | ||
| D38082.1 | - | 7 | 927 | |||
| S75359.1 | - | 8 | 1070 | |||