siRNA Details
| Gene Symbol: Marcksl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030862.2 | NP_110489.1 | 2 | 594 | ||
| GenBank mRNA | AJ301677.1 | - | 2 | 487 | ||
| BC081789.1 | - | 2 | 594 | 7134056 | ||
| Gene Symbol: LOC102550530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759283.1 | XP_008757505.1 | 2 | 553 | ||

Marcksl1
Genomic Map