siRNA Details
| Gene Symbol: Adar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031006.1 | NP_112268.1 | 8 | 2459 | ||
| XM_006232774.2 | XP_006232836.1 | 9 | 2683 | |||
| XM_006232775.2 | XP_006232837.1 | 9 | 2604 | |||
| XM_006232776.2 | XP_006232838.1 | 8 | 5567 | |||
| XM_006232777.2 | XP_006232839.1 | 8 | 2491 | |||
| XM_006232778.2 | XP_006232840.1 | 8 | 2443 | |||
| GenBank mRNA | FQ212262.1 | - | 2667 | |||
| FQ212270.1 | - | 2667 | ||||
| U18942.1 | - | 8 | 2459 | |||

Adar
Genomic Map