siRNA Details
| Gene Symbol: Adcy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031007.1 | NP_112269.1 | 13 | 1898 | ||
| GenBank mRNA | M80550.1 | - | 13 | 1898 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adcy2| Gene Symbol: Adcy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031007.1 | NP_112269.1 | 13 | 1898 | ||
| GenBank mRNA | M80550.1 | - | 13 | 1898 | ||