siRNA Details
Gene Symbol: Abcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031013.1 | NP_112275.1 | 12 | 1683 | ||
XM_006229159.2 | XP_006229221.1 | 12 | 1600 | |||
XM_008759419.1 | XP_008757641.1 | 11 | 1929 | |||
GenBank mRNA | AB010466.1 | - | 12 | 1636 | ||
U73038.1 | - | 12 | 1683 |
